Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLLT10
Entrez Gene ID
8028
Gene Name
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 10
Gene Aliases
AF10
Chromosome Location
Chr.10: 21533349 - 21743630 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195626.1 NP_001182555.1 14 - 15 1989 134
NM_001324297.1 NP_001311226.1 17 - 18 2822 134
NM_004641.3 NP_004632.1 16 - 17 2386 134
NR_136736.1 18 - 19 2634 134
GenBank mRNA AB209755.1 - 18 - 19 2340 134
AY598745.1 - 14 - 15 1989 134
BC129946.1 - 15 - 16 2032 134
BX640774.1 - 4 - 5 587 134
BX648210.1 - 8 - 9 1074 134
U13948.1 - 16 - 17 2220 134

View Full Product Details