The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204514.1 | NP_001191443.1 | 16 - 17 | 2237 | 98 |
NM_001204516.1 | NP_001191445.1 | 14 - 15 | 1941 | 98 | |
NM_001204517.1 | NP_001191446.1 | 15 - 16 | 3182 | 98 | |
NM_014970.3 | NP_055785.2 | 15 - 16 | 2073 | 98 | |
XM_005244970.1 | XP_005245027.1 | 15 - 16 | 2076 | 98 | |
XM_011509307.2 | XP_011507609.2 | 15 - 16 | 2076 | 98 | |
GenBank mRNA | AK026471.1 | - | 15 - 16 | 1852 | 98 |
AK122897.1 | - | 15 - 16 | 3184 | 98 | |
AK297368.1 | - | 14 - 15 | 1868 | 98 | |
AK301912.1 | - | 8 - 9 | 1040 | 98 | |
AK303052.1 | - | 16 - 17 | 2194 | 98 | |
AK315586.1 | - | 15 - 16 | 1987 | 98 | |
BC028679.2 | - | 15 - 16 | 2030 | 98 | |
BC048012.1 | - | 14 - 15 | 1985 | 98 | |
U59919.1 | - | 15 - 16 | 1990 | 98 |