Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLLT10
Entrez Gene ID
8028
Gene Name
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 10
Gene Aliases
AF10
Chromosome Location
Chr.10: 21533349 - 21743630 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195626.1 NP_001182555.1 7 - 8 688 195
NM_001324297.1 NP_001311226.1 9 - 10 1385 195
NM_004641.3 NP_004632.1 8 - 9 1037 195
NR_136736.1 9 - 10 1215 195
GenBank mRNA AB209755.1 - 9 - 10 921 195
AY598745.1 - 7 - 8 688 195
U13948.1 - 8 - 9 871 195

View Full Product Details