The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153690.4 | NP_710157.2 | 1 - 1 | 3024 | 137 |
GenBank mRNA | BI494721.1 | - | 1 - 1 | 233 | 137 |