The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033123.3 | NP_149114.2 | 8 - 9 | 1218 | 142 |
NR_073075.1 | 8 - 9 | 1221 | 142 | ||
XM_011520883.1 | XP_011519185.1 | 5 - 6 | 624 | 142 | |
XM_017020177.1 | XP_016875666.1 | 8 - 9 | 1211 | 142 | |
XM_017020186.1 | XP_016875675.1 | 5 - 6 | 586 | 142 | |
GenBank mRNA | AF532185.1 | - | 8 - 9 | 1143 | 142 |
AK097555.1 | - | 4 - 5 | 566 | 142 | |
AK292279.1 | - | 8 - 9 | 1216 | 142 | |
AK314516.1 | - | 8 - 9 | 1175 | 142 | |
AY035866.1 | - | 8 - 9 | 1180 | 142 | |
BC125067.1 | - | 4 - 5 | 471 | 142 |