The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317997.1 | NP_001304926.1 | 2 - 3 | 181 | 71 |
NM_032307.4 | NP_115683.3 | 1 - 2 | 579 | 71 | |
XM_005252271.4 | XP_005252328.1 | 1 - 2 | 580 | 71 | |
GenBank mRNA | AK090882.1 | - | 2 - 3 | 181 | 71 |
BC004407.1 | - | 1 - 2 | 554 | 71 | |
BC137461.1 | - | 1 - 2 | 456 | 71 | |
BC137462.1 | - | 1 - 2 | 456 | 71 |