The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000938.2 | NP_000929.1 | 22 - 23 | 3424 | 97 |
NM_001303268.1 | NP_001290197.1 | 21 - 22 | 3273 | 97 | |
NM_001303269.1 | NP_001290198.1 | 23 - 24 | 3602 | 97 | |
GenBank mRNA | AF055028.1 | - | 8 - 9 | 984 | 97 |
AK289823.1 | - | 22 - 23 | 3168 | 97 | |
AK294901.1 | - | 21 - 22 | 3049 | 97 | |
AK295132.1 | - | 23 - 24 | 3373 | 97 | |
AK299090.1 | - | 22 - 23 | 2940 | 97 | |
BC023503.2 | - | 22 - 23 | 3182 | 97 | |
BU619780.1 | - | 3 - 4 | 97 | ||
X63563.1 | - | 22 - 23 | 3121 | 97 |