The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004523.3 | NP_004514.2 | 17 - 18 | 2567 | 146 |
GenBank mRNA | AB208964.1 | - | 3 - 4 | 302 | 146 |
AK314258.1 | - | 17 - 18 | 2470 | 146 | |
BC126211.1 | - | 17 - 18 | 2366 | 146 | |
BC136474.1 | - | 17 - 18 | 2366 | 146 | |
U37426.1 | - | 17 - 18 | 2349 | 146 | |
X85137.1 | - | 17 - 18 | 2272 | 146 |