The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004523.3 | NP_004514.2 | 1 - 2 | 385 | 96 |
GenBank mRNA | AA857025.1 | - | 1 - 2 | 217 | 95 |
AK314258.1 | - | 1 - 2 | 288 | 96 | |
BC126211.1 | - | 1 - 2 | 184 | 96 | |
BC136474.1 | - | 1 - 2 | 184 | 96 | |
DB078134.1 | - | 1 - 2 | 385 | 96 | |
U37426.1 | - | 1 - 2 | 167 | 96 | |
X85137.1 | - | 1 - 2 | 87 | 96 |