Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LIG3
Entrez Gene ID
3980
Gene Name
DNA ligase 3
Gene Aliases
LIG2
Chromosome Location
Chr.17: 34980498 - 35009169 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002311.4 NP_002302.2 17 - 18 2609 87
NM_013975.3 NP_039269.2 17 - 18 2609 87
XM_005257970.3 XP_005258027.1 17 - 18 2577 87
XM_006721896.3 XP_006721959.1 17 - 18 2578 87
XM_017024624.1 XP_016880113.1 17 - 18 2773 87
GenBank mRNA AK125853.1 - 4 - 5 2333 87
AK300363.1 - 17 - 18 2569 87
BC068005.1 - 17 - 18 2584 87
BG470103.1 - 1 - 2 92 87
U40671.1 - 17 - 18 2570 87
X84740.1 - 17 - 18 2552 87

View Full Product Details