The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128844.1 | NP_001122316.1 | 19 - 20 | 2976 | 74 |
NM_001128845.1 | NP_001122317.1 | 17 - 18 | 2616 | 74 | |
NM_001128846.1 | NP_001122318.1 | 17 - 18 | 2616 | 74 | |
NM_001128847.1 | NP_001122319.1 | 17 - 18 | 2616 | 74 | |
NM_001128848.1 | NP_001122320.1 | 17 - 18 | 2616 | 74 | |
NM_001128849.1 | NP_001122321.1 | 18 - 19 | 2690 | 74 | |
NM_003072.3 | NP_003063.2 | 18 - 19 | 2900 | 74 | |
XM_006722845.2 | XP_006722908.1 | 18 - 19 | 2822 | 74 | |
XM_006722846.2 | XP_006722909.1 | 18 - 19 | 2916 | 74 | |
XM_011528198.1 | XP_011526500.1 | 19 - 20 | 2897 | 74 | |
XM_017027160.1 | XP_016882649.1 | 19 - 20 | 2897 | 74 | |
XM_017027161.1 | XP_016882650.1 | 19 - 20 | 2897 | 74 | |
XM_017027162.1 | XP_016882651.1 | 19 - 20 | 2897 | 74 | |
XM_017027163.1 | XP_016882652.1 | 19 - 20 | 2897 | 74 | |
XM_017027164.1 | XP_016882653.1 | 19 - 20 | 2897 | 74 | |
XM_017027165.1 | XP_016882654.1 | 19 - 20 | 2897 | 74 | |
XM_017027166.1 | XP_016882655.1 | 19 - 20 | 2897 | 74 | |
XM_017027167.1 | XP_016882656.1 | 19 - 20 | 2897 | 74 | |
XM_017027168.1 | XP_016882657.1 | 19 - 20 | 2897 | 74 | |
GenBank mRNA | AB209313.2 | - | 17 - 18 | 2490 | 74 |
AK055168.1 | - | 1 - 2 | 688 | 74 | |
AK290033.1 | - | 19 - 20 | 2897 | 74 | |
AK299761.1 | - | 4 - 5 | 358 | 74 | |
AK303352.1 | - | 3 - 4 | 300 | 74 | |
AK308763.1 | - | 3 - 4 | 529 | 74 | |
BC136644.1 | - | 16 - 17 | 2836 | 74 | |
BC150298.1 | - | 18 - 19 | 2821 | 74 | |
D26156.1 | - | 18 - 19 | 2673 | 74 | |
EU430756.1 | - | 17 - 18 | 2616 | 74 | |
EU430757.1 | - | 17 - 18 | 2616 | 74 | |
EU430758.1 | - | 17 - 18 | 2616 | 74 | |
EU430759.1 | - | 17 - 18 | 2616 | 74 | |
U29175.1 | - | 18 - 19 | 2694 | 74 |