Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMARCA4
Entrez Gene ID
6597
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4
Gene Aliases
BAF190, BAF190A, BRG1, CSS4, MRD16, RTPS2, SNF2, SNF2L4, SNF2LB, SWI2, hSNF2b
Chromosome Location
Chr.19: 10960922 - 11062282 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128844.1 NP_001122316.1 27 - 28 4134 90
NM_001128849.1 NP_001122321.1 26 - 27 3848 90
NM_003072.3 NP_003063.2 26 - 27 4058 90
XM_006722845.2 XP_006722908.1 26 - 27 3980 90
XM_006722846.2 XP_006722909.1 26 - 27 4074 90
XM_011528198.1 XP_011526500.1 27 - 28 4055 90
XM_017027160.1 XP_016882649.1 27 - 28 4055 90
XM_017027161.1 XP_016882650.1 27 - 28 4055 90
XM_017027163.1 XP_016882652.1 27 - 28 4055 90
GenBank mRNA AK290033.1 - 27 - 28 4055 90
AK299761.1 - 12 - 13 1516 90
BC150298.1 - 26 - 27 3979 90
D26156.1 - 26 - 27 3831 90
U29175.1 - 26 - 27 3852 90

View Full Product Details