Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGEF15
Entrez Gene ID
22899
Gene Name
Rho guanine nucleotide exchange factor 15
Gene Aliases
ARGEF15, E5, Ephexin5, Vsm-RhoGEF
Chromosome Location
Chr.17: 8310238 - 8322516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025014.1 NP_079290.1 2 - 3 688 80
NM_173728.3 NP_776089.2 2 - 3 752 80
XM_011523734.1 XP_011522036.1 2 - 3 752 80
XM_011523735.1 XP_011522037.1 2 - 3 688 80
XM_011523736.1 XP_011522038.1 2 - 3 752 80
GenBank mRNA AB020722.1 - 2 - 3 668 80
AK291626.1 - 2 - 3 712 80
BC036749.1 - 2 - 3 707 80
KU178557.1 - 1 - 2 597 80
KU178558.1 - 1 - 2 597 80

View Full Product Details