Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRS2
Entrez Gene ID
57380
Gene Name
MRS2, magnesium transporter
Gene Aliases
HPT, MRS2L
Chromosome Location
Chr.6: 24402908 - 24426194 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_104423.1 4 - 5 650 100
GenBank mRNA AK297418.1 - 4 - 5 627 100

View Full Product Details