The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024307.2 | NP_077283.2 | 2 - 3 | 264 | 95 |
GenBank mRNA | BC002714.2 | - | 2 - 3 | 264 | 95 |