Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FLYWCH1
Entrez Gene ID
84256
Gene Name
FLYWCH-type zinc finger 1
Gene Aliases
-
Chromosome Location
Chr.16: 2911873 - 2951208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308068.1 NP_001294997.1 2 - 3 296 158
NM_020912.1 NP_065963.1 2 - 3 296 158
NM_032296.2 NP_115672.2 2 - 3 296 158
XM_006720962.3 XP_006721025.1 2 - 3 398 158
XM_011522701.2 XP_011521003.1 2 - 3 400 158
XM_011522702.2 XP_011521004.1 2 - 3 874 158
XM_011522704.2 XP_011521006.1 2 - 3 401 158
XM_011522705.2 XP_011521007.1 2 - 3 399 158
XM_011522707.2 XP_011521009.1 2 - 3 402 158
GenBank mRNA AB046772.1 - 2 - 3 276 158
BC028572.1 - 2 - 3 297 158
CN359886.1 - 2 - 3 335 158

View Full Product Details