Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLYWCH1
Entrez Gene ID
84256
Gene Name
FLYWCH-type zinc finger 1
Gene Aliases
-
Chromosome Location
Chr.16: 2911873 - 2951208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308068.1 NP_001294997.1 6 - 7 1883 93
NM_020912.1 NP_065963.1 6 - 7 1880 93
NM_032296.2 NP_115672.2 6 - 7 1880 93
GenBank mRNA AB046772.1 - 6 - 7 1860 93
AK026885.1 - 1 - 2 49 93
AK092434.1 - 2 - 3 700 93
BC001973.1 - 3 - 4 808 93
BC018712.1 - 2 - 3 628 93
BC028572.1 - 6 - 7 1884 93

View Full Product Details