The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253725.1 | NP_001240654.1 | 3 - 4 | 668 | 67 |
NM_001253726.1 | NP_001240655.1 | 3 - 4 | 668 | 67 | |
NM_024507.3 | NP_078783.1 | 3 - 4 | 668 | 67 | |
NM_172229.2 | NP_757384.1 | 3 - 4 | 668 | 67 | |
GenBank mRNA | AB086355.1 | - | 3 - 4 | 363 | 67 |
AB086356.1 | - | 3 - 4 | 363 | 67 | |
AB086357.1 | - | 3 - 4 | 363 | 67 | |
AB086405.1 | - | 3 - 4 | 363 | 67 | |
AK027669.1 | - | 3 - 4 | 572 | 67 | |
AK075033.1 | - | 3 - 4 | 609 | 67 | |
AK223422.1 | - | 3 - 4 | 440 | 67 | |
AK301953.1 | - | 3 - 4 | 609 | 67 | |
BC003533.1 | - | 3 - 4 | 475 | 67 | |
BC009383.2 | - | 3 - 4 | 668 | 67 |