The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018037.1 | NP_001018047.1 | 37 - 38 | 4777 | 141 |
NM_001018038.2 | NP_001018048.1 | 38 - 39 | 4894 | 141 | |
NM_015186.3 | NP_056001.1 | 38 - 39 | 4894 | 141 | |
NM_033305.2 | NP_150648.2 | 38 - 39 | 4894 | 141 | |
GenBank mRNA | AB054005.1 | - | 38 - 39 | 4805 | 141 |
AF337532.1 | - | 38 - 39 | 4894 | 141 | |
AJ608769.1 | - | 37 - 38 | 4777 | 141 | |
AJ626859.1 | - | 38 - 39 | 4894 | 141 | |
AK000273.1 | - | 8 - 9 | 1223 | 141 | |
AK124024.1 | - | 13 - 14 | 1924 | 141 |