The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018037.1 | NP_001018047.1 | 52 - 53 | 7561 | 98 |
NM_001018038.2 | NP_001018048.1 | 53 - 54 | 7678 | 98 | |
NM_015186.3 | NP_056001.1 | 53 - 54 | 7678 | 98 | |
NM_033305.2 | NP_150648.2 | 53 - 54 | 7678 | 98 | |
GenBank mRNA | AB023203.1 | - | 14 - 15 | 2509 | 98 |
AB054005.1 | - | 53 - 54 | 7589 | 98 | |
AF337532.1 | - | 53 - 54 | 7678 | 98 | |
AJ608769.1 | - | 52 - 53 | 7561 | 98 | |
AJ626859.1 | - | 53 - 54 | 7678 | 98 | |
AK022967.1 | - | 6 - 7 | 836 | 98 | |
BC041852.1 | - | 11 - 12 | 1873 | 98 |