The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020310.2 | NP_064706.1 | 4 - 5 | 1056 | 101 |
XM_011523868.2 | XP_011522170.1 | 4 - 5 | 993 | 101 | |
GenBank mRNA | AK291596.1 | - | 4 - 5 | 1153 | 101 |
BC117563.1 | - | 4 - 5 | 806 | 101 | |
X96401.1 | - | 4 - 5 | 1016 | 101 |