Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLXNA2
Entrez Gene ID
5362
Gene Name
plexin A2
Gene Aliases
OCT, PLXN2
Chromosome Location
Chr.1: 208022242 - 208244384 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025179.3 NP_079455.3 14 - 15 3613 64
XM_005273164.2 XP_005273221.1 14 - 15 3677 64
XM_005273165.3 XP_005273222.1 14 - 15 3677 64
GenBank mRNA AB007932.1 - 14 - 15 3064 64
BC132676.1 - 14 - 15 2968 64
BC136530.1 - 14 - 15 2968 64
X87831.2 - 12 - 13 1518 64

View Full Product Details