The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000356.3 | NP_000347.2 | 19 - 20 | 3179 | 60 |
NM_001135243.1 | NP_001128715.1 | 20 - 21 | 3410 | 60 | |
NM_001135245.1 | NP_001128717.1 | 19 - 20 | 3179 | 60 | |
XM_005268502.3 | XP_005268559.1 | 21 - 22 | 3458 | 60 | |
XM_005268503.3 | XP_005268560.1 | 21 - 22 | 3455 | 60 | |
XM_005268504.3 | XP_005268561.1 | 21 - 22 | 3458 | 60 | |
XM_005268505.3 | XP_005268562.1 | 20 - 21 | 3347 | 60 | |
XM_005268507.3 | XP_005268564.1 | 20 - 21 | 3227 | 60 | |
XM_011537678.2 | XP_011535980.1 | 20 - 21 | 3278 | 60 | |
XM_017009793.1 | XP_016865282.1 | 19 - 20 | 3167 | 60 | |
GenBank mRNA | AK096467.1 | - | 16 - 17 | 2915 | 60 |
AK307586.1 | - | 20 - 21 | 3335 | 60 | |
AY460334.1 | - | 20 - 21 | 3302 | 60 | |
BC008331.1 | - | 11 - 12 | 1976 | 60 | |
BC027252.1 | - | 19 - 20 | 3027 | 60 | |
U40847.1 | - | 19 - 20 | 3164 | 60 | |
U76366.1 | - | 19 - 20 | 3146 | 60 |