The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005963.3 | NP_005954.3 | 21 - 22 | 2523 | 68 |
XM_017024675.1 | XP_016880164.1 | 21 - 22 | 2696 | 68 | |
GenBank mRNA | AF111785.1 | - | 19 - 20 | 2429 | 68 |
BC114545.1 | - | 19 - 20 | 2429 | 68 | |
EU794637.1 | - | 19 - 20 | 2429 | 68 |