Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DYNLT3
Entrez Gene ID
6990
Gene Name
dynein light chain Tctex-type 3
Gene Aliases
RP3, TCTE1L, TCTEX1L
Chromosome Location
Chr.X: 37838836 - 37847636 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006520.2 NP_006511.1 4 - 5 398 85
GenBank mRNA AK311851.1 - 4 - 5 333 85
BC000968.2 - 4 - 5 326 85
CR450320.1 - 4 - 5 272 85
CR542213.1 - 4 - 5 272 85
DA342302.1 - 4 - 5 389 85
U02556.1 - 4 - 5 340 85

View Full Product Details