Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYTL5
Entrez Gene ID
94122
Gene Name
synaptotagmin like 5
Gene Aliases
slp5
Chromosome Location
Chr.X: 37888915 - 38128820 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163334.1 NP_001156806.1 7 - 8 1313 130
NM_001163335.1 NP_001156807.1 9 - 10 1491 130
NM_138780.2 NP_620135.1 8 - 9 1400 130
XM_011544001.2 XP_011542303.1 10 - 11 1752 130
XM_011544002.2 XP_011542304.1 8 - 9 1386 130
XM_017029972.1 XP_016885461.1 11 - 12 1918 130
XM_017029973.1 XP_016885462.1 10 - 11 1985 130
XM_017029974.1 XP_016885463.1 9 - 10 1613 130
GenBank mRNA AB080222.1 - 7 - 8 957 130
AK125106.1 - 8 - 9 1368 130
BC131585.1 - 7 - 8 957 130
BX647688.1 - 9 - 10 1493 130

View Full Product Details