The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319193.1 | NP_001306122.1 | 25 - 26 | 3076 | 97 |
GenBank mRNA | AB354594.1 | - | 24 - 25 | 2993 | 97 |
AK057261.1 | - | 7 - 8 | 894 | 97 | |
AK074045.1 | - | 24 - 25 | 3148 | 97 | |
BC007570.1 | - | 12 - 13 | 1496 | 97 |