The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319193.1 | NP_001306122.1 | 12 - 13 | 1051 | 79 |
GenBank mRNA | AB354594.1 | - | 11 - 12 | 968 | 79 |
AK074045.1 | - | 12 - 13 | 1046 | 79 | |
BC023549.2 | - | 12 - 13 | 1187 | 79 |