The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000296.3 | NP_000287.3 | 35 - 36 | 10830 | 160 |
NM_001009944.2 | NP_001009944.2 | 35 - 36 | 10833 | 160 | |
XM_005255370.3 | XP_005255427.1 | 25 - 26 | 8420 | 160 | |
XM_011522528.2 | XP_011520830.1 | 36 - 37 | 10858 | 160 | |
XM_011522529.2 | XP_011520831.1 | 36 - 37 | 10856 | 160 | |
XM_011522530.2 | XP_011520832.1 | 36 - 37 | 10663 | 160 | |
XM_011522532.2 | XP_011520834.1 | 35 - 36 | 10591 | 160 | |
XM_011522534.2 | XP_011520836.1 | 36 - 37 | 10648 | 160 | |
XM_011522535.2 | XP_011520837.1 | 27 - 28 | 8880 | 160 | |
XM_011522537.2 | XP_011520839.1 | 25 - 26 | 7791 | 160 | |
XM_017023285.1 | XP_016878774.1 | 26 - 27 | 8435 | 160 | |
GenBank mRNA | AB209025.1 | - | 22 - 23 | 5187 | 160 |
AB209675.1 | - | 21 - 22 | 6139 | 160 | |
L33243.1 | - | 35 - 36 | 10832 | 160 | |
U24497.1 | - | 35 - 36 | 10759 | 160 |