The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317237.1 | NP_001304166.1 | 9 - 10 | 1066 | 81 |
NM_001908.4 | NP_001899.1 | 9 - 10 | 1097 | 81 | |
NM_147780.3 | NP_680090.1 | 11 - 12 | 1259 | 81 | |
NM_147781.3 | NP_680091.1 | 10 - 11 | 1216 | 81 | |
NM_147782.3 | NP_680092.1 | 10 - 11 | 1185 | 81 | |
NM_147783.3 | NP_680093.1 | 10 - 11 | 1171 | 81 | |
XM_006716244.2 | XP_006716307.1 | 11 - 12 | 1283 | 81 | |
XM_006716245.2 | XP_006716308.1 | 9 - 10 | 1387 | 81 | |
XM_011543812.2 | XP_011542114.1 | 11 - 12 | 1192 | 81 | |
XM_017013097.1 | XP_016868586.1 | 10 - 11 | 1173 | 81 | |
XM_017013098.1 | XP_016868587.1 | 10 - 11 | 1173 | 81 | |
XM_017013099.1 | XP_016868588.1 | 11 - 12 | 1215 | 81 | |
XM_017013100.1 | XP_016868589.1 | 10 - 11 | 1117 | 81 | |
XM_017013101.1 | XP_016868590.1 | 6 - 7 | 640 | 81 | |
GenBank mRNA | AK075393.1 | - | 9 - 10 | 1073 | 81 |
AK092070.1 | - | 10 - 11 | 1108 | 81 | |
AK097384.1 | - | 4 - 5 | 1027 | 81 | |
AK130184.1 | - | 9 - 10 | 1034 | 81 | |
AK290239.1 | - | 9 - 10 | 1034 | 81 | |
AK296845.1 | - | 8 - 9 | 838 | 81 | |
AK297687.1 | - | 7 - 8 | 798 | 81 | |
BC010240.1 | - | 10 - 11 | 1114 | 81 | |
BC095408.1 | - | 9 - 10 | 1034 | 81 | |
BX647765.1 | - | 9 - 10 | 957 | 81 | |
BX647952.1 | - | 9 - 10 | 1037 | 81 | |
L16510.1 | - | 10 - 11 | 1099 | 81 | |
M13230.1 | - | 5 - 6 | 532 | 81 |