Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POF1B
Entrez Gene ID
79983
Gene Name
premature ovarian failure, 1B
Gene Aliases
POF, POF2B
Chromosome Location
Chr.X: 85277389 - 85379743 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307940.1 NP_001294869.1 12 - 13 1463 144
NM_024921.3 NP_079197.3 12 - 13 1463 144
XM_005262203.3 XP_005262260.1 12 - 13 1376 144
GenBank mRNA AF309774.1 - 12 - 13 1463 144
AK025039.1 - 8 - 9 1056 144
AK025080.1 - 4 - 5 428 144
AK225193.1 - 4 - 5 428 144
AK290360.1 - 12 - 13 1412 144
AK314295.1 - 12 - 13 1382 144
BC017500.1 - 12 - 13 1437 144

View Full Product Details