Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POF1B
Entrez Gene ID
79983
Gene Name
premature ovarian failure, 1B
Gene Aliases
POF, POF2B
Chromosome Location
Chr.X: 85277389 - 85379743 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307940.1 NP_001294869.1 3 - 4 499 133
NM_024921.3 NP_079197.3 3 - 4 499 133
XM_005262203.3 XP_005262260.1 3 - 4 457 133
GenBank mRNA AF309774.1 - 3 - 4 499 133
AK290360.1 - 3 - 4 448 133
AK314295.1 - 3 - 4 418 133
BC017500.1 - 3 - 4 473 133

View Full Product Details