The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001502.3 | NP_001493.2 | 3 - 4 | 723 | 63 |
GenBank mRNA | AB035541.1 | - | 2 - 3 | 512 | 63 |
AK312277.1 | - | 3 - 4 | 610 | 63 | |
BX648023.1 | - | 3 - 4 | 617 | 63 | |
CR749600.1 | - | 3 - 4 | 682 | 63 | |
U36221.1 | - | 3 - 4 | 592 | 63 |