Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CELSR1
Entrez Gene ID
9620
Gene Name
cadherin EGF LAG seven-pass G-type receptor 1
Gene Aliases
ADGRC1, CDHF9, FMI2, HFMI2, ME2
Chromosome Location
Chr.22: 46360834 - 46537502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014246.1 NP_055061.1 14 - 15 5964 98
XM_006724383.3 XP_006724446.1 14 - 15 6291 98
XM_011530553.1 XP_011528855.1 14 - 15 6296 98
XM_011530554.2 XP_011528856.1 14 - 15 2507 98
XM_011530555.2 XP_011528857.1 14 - 15 2588 98
GenBank mRNA AF231024.1 - 14 - 15 5964 98
AK302517.1 - 6 - 7 1042 98

View Full Product Details