Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEK10
Entrez Gene ID
152110
Gene Name
NIMA related kinase 10
Gene Aliases
-
Chromosome Location
Chr.3: 27107336 - 27369460 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152534.4 NP_689747.3 5 - 6 577 85
NM_199347.3 NP_955379.2 5 - 6 577 85
XM_006712997.3 XP_006713060.1 5 - 6 538 85
XM_006712998.2 XP_006713061.1 5 - 6 538 85
XM_006712999.3 XP_006713062.1 5 - 6 538 85
XM_006713001.3 XP_006713064.1 5 - 6 538 85
XM_006713002.1 XP_006713065.1 5 - 6 538 85
XM_011533413.2 XP_011531715.1 5 - 6 538 85
XM_011533414.2 XP_011531716.1 5 - 6 538 85
XM_011533415.1 XP_011531717.1 5 - 6 538 85
XM_017005761.1 XP_016861250.1 4 - 5 490 85
XM_017005762.1 XP_016861251.1 6 - 7 654 85
XM_017005763.1 XP_016861252.1 5 - 6 635 85
XM_017005764.1 XP_016861253.1 4 - 5 467 85
XM_017005765.1 XP_016861254.1 4 - 5 439 85
XM_017005766.1 XP_016861255.1 4 - 5 490 85
XM_017005767.1 XP_016861256.1 4 - 5 490 85
XM_017005768.1 XP_016861257.1 5 - 6 538 85
XM_017005769.1 XP_016861258.1 5 - 6 538 85
XM_017005770.1 XP_016861259.1 4 - 5 490 85
XM_017005771.1 XP_016861260.1 4 - 5 490 85
XM_017005772.1 XP_016861261.1 4 - 5 490 85
XM_017005773.1 XP_016861262.1 5 - 6 538 85
XM_017005774.1 XP_016861263.1 4 - 5 490 85
GenBank mRNA AK123061.1 - 5 - 6 538 85
AK128585.1 - 4 - 5 467 85
BX537852.1 - 1 - 2 44 85

View Full Product Details