The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AJ276510.1 | - | 1 - 1 | 58 | 97 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | ATPase phospholipid transporting 9B (putative) | ATP9B | ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1 | -137.5kb | Anti-sense | Chr. - 79069275 - 79378391 on Build GRCh38 |
Downstream | uncharacterized LOC284240 | LOC284240 | 58.6kb | Anti-sense | Chr. - 79573705 - 79591904 on Build GRCh38 |