The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001789.2 | NP_001780.2 | 13 - 14 | 1691 | 68 |
NM_201567.1 | NP_963861.1 | 12 - 13 | 1571 | 68 | |
XM_006713434.2 | XP_006713497.1 | 13 - 14 | 1783 | 68 | |
XM_006713435.2 | XP_006713498.1 | 12 - 13 | 1663 | 68 | |
XM_011534316.1 | XP_011532618.1 | 14 - 15 | 1884 | 68 | |
GenBank mRNA | AK290554.1 | - | 13 - 14 | 1366 | 68 |
AY137580.1 | - | 12 - 13 | 1207 | 68 | |
BC007401.2 | - | 13 - 14 | 1691 | 68 | |
BC018642.2 | - | 13 - 14 | 1691 | 68 | |
EU812118.1 | - | 12 - 13 | 1204 | 68 | |
FJ866789.1 | - | 13 - 14 | 1324 | 68 | |
M81933.1 | - | 13 - 14 | 1783 | 68 |