The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111034.2 | NP_001104504.1 | 1 - 2 | 195 | 138 |
NM_001111035.2 | NP_001104505.1 | 2 - 3 | 237 | 138 | |
XM_005259938.1 | XP_005259995.1 | 2 - 3 | 315 | 138 | |
GenBank mRNA | AK225912.1 | - | 1 - 2 | 192 | 138 |
AK307164.1 | - | 2 - 3 | 205 | 138 | |
BC111014.1 | - | 2 - 3 | 180 | 138 | |
BP367008.1 | - | 1 - 2 | 198 | 138 |