Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLF16
Entrez Gene ID
83855
Gene Name
Kruppel like factor 16
Gene Aliases
BTEB4, DRRF, NSLP2
Chromosome Location
Chr.19: 1852399 - 1863565 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF327440.1 - 2 - 3 941 118

View Full Product Details