Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FHAD1
- Entrez Gene ID
-
114827
- Gene Name
- forkhead associated phosphopeptide binding domain 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 15236559 - 15400283 on Build GRCh38
View Full Product Details