Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FHAD1
Entrez Gene ID
114827
Gene Name
forkhead associated phosphopeptide binding domain 1
Gene Aliases
-
Chromosome Location
Chr.1: 15236559 - 15400283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052929.1 NP_443161.1 17 - 18 2417 113
XM_011540575.2 XP_011538877.1 18 - 19 2559 113
XM_011540576.2 XP_011538878.1 18 - 19 2560 113
XM_011540577.2 XP_011538879.1 18 - 19 2521 113
XM_011540580.2 XP_011538882.1 18 - 19 2558 113
XM_011540581.2 XP_011538883.1 18 - 19 2558 113
XM_011540582.2 XP_011538884.1 18 - 19 2558 113
XM_011540584.2 XP_011538886.1 18 - 19 2559 113
XM_011540588.2 XP_011538890.1 18 - 19 2550 113
XM_011540590.2 XP_011538892.1 18 - 19 2551 113
XM_011540592.2 XP_011538894.1 17 - 18 2260 113
XM_011540595.2 XP_011538897.1 15 - 16 1903 113
XM_011540596.2 XP_011538898.1 12 - 13 1463 113
XM_011540597.2 XP_011538899.1 18 - 19 2560 113
XM_011540598.2 XP_011538900.1 17 - 18 2491 113
XM_017000198.1 XP_016855687.1 18 - 19 2560 113
XM_017000199.1 XP_016855688.1 18 - 19 2560 113
XM_017000200.1 XP_016855689.1 18 - 19 2559 113
XM_017000201.1 XP_016855690.1 18 - 19 2559 113
XM_017000202.1 XP_016855691.1 17 - 18 2452 113
XM_017000203.1 XP_016855692.1 18 - 19 2556 113
XM_017000204.1 XP_016855693.1 18 - 19 2555 113
XM_017000205.1 XP_016855694.1 16 - 17 2365 113
XM_017000206.1 XP_016855695.1 18 - 19 2555 113
XM_017000207.1 XP_016855696.1 17 - 18 2324 113
XM_017000208.1 XP_016855697.1 18 - 19 2550 113
XM_017000209.1 XP_016855698.1 18 - 19 2554 113
XM_017000210.1 XP_016855699.1 18 - 19 2550 113
XM_017000212.1 XP_016855701.1 18 - 19 2549 113
XM_017000213.1 XP_016855702.1 18 - 19 2559 113
XM_017000214.1 XP_016855703.1 18 - 19 2560 113
GenBank mRNA AL833342.1 - 7 - 8 816 113

View Full Product Details