Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FHAD1
Entrez Gene ID
114827
Gene Name
forkhead associated phosphopeptide binding domain 1
Gene Aliases
-
Chromosome Location
Chr.1: 15236559 - 15400283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052929.1 NP_443161.1 9 - 10 1400 81
XM_011540575.2 XP_011538877.1 9 - 10 1476 81
XM_011540576.2 XP_011538878.1 9 - 10 1477 81
XM_011540577.2 XP_011538879.1 9 - 10 1438 81
XM_011540580.2 XP_011538882.1 9 - 10 1475 81
XM_011540581.2 XP_011538883.1 9 - 10 1475 81
XM_011540582.2 XP_011538884.1 9 - 10 1475 81
XM_011540584.2 XP_011538886.1 9 - 10 1476 81
XM_011540588.2 XP_011538890.1 9 - 10 1467 81
XM_011540589.2 XP_011538891.1 9 - 10 1476 81
XM_011540590.2 XP_011538892.1 9 - 10 1468 81
XM_011540592.2 XP_011538894.1 8 - 9 1177 81
XM_011540593.2 XP_011538895.1 9 - 10 1472 81
XM_011540595.2 XP_011538897.1 6 - 7 820 81
XM_011540596.2 XP_011538898.1 3 - 4 380 81
XM_011540597.2 XP_011538899.1 9 - 10 1477 81
XM_011540598.2 XP_011538900.1 9 - 10 1479 81
XM_017000198.1 XP_016855687.1 9 - 10 1477 81
XM_017000199.1 XP_016855688.1 9 - 10 1477 81
XM_017000200.1 XP_016855689.1 9 - 10 1476 81
XM_017000201.1 XP_016855690.1 9 - 10 1476 81
XM_017000202.1 XP_016855691.1 9 - 10 1477 81
XM_017000203.1 XP_016855692.1 9 - 10 1473 81
XM_017000204.1 XP_016855693.1 9 - 10 1472 81
XM_017000206.1 XP_016855695.1 9 - 10 1472 81
XM_017000207.1 XP_016855696.1 8 - 9 1241 81
XM_017000208.1 XP_016855697.1 9 - 10 1467 81
XM_017000209.1 XP_016855698.1 9 - 10 1471 81
XM_017000210.1 XP_016855699.1 9 - 10 1467 81
XM_017000211.1 XP_016855700.1 9 - 10 1475 81
XM_017000212.1 XP_016855701.1 9 - 10 1466 81
XM_017000213.1 XP_016855702.1 9 - 10 1476 81
XM_017000214.1 XP_016855703.1 9 - 10 1477 81
GenBank mRNA AK057963.1 - 8 - 9 1218 81
AK093300.1 - 4 - 5 372 81

View Full Product Details