Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNF26
Entrez Gene ID
7574
Gene Name
zinc finger protein 26
Gene Aliases
HEL-179, KOX20
Chromosome Location
Chr.12: 132986348 - 133027102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256279.1 NP_001243208.1 5 - 5 3148 121
NM_001256280.1 NP_001243209.1 3 - 3 2953 121
NM_019591.3 NP_062537.2 4 - 4 3049 121
XM_005266182.3 XP_005266239.1 5 - 5 3136 121
XM_005266183.3 XP_005266240.1 6 - 6 3339 121
XM_011534829.1 XP_011533131.1 5 - 5 3252 121
XM_017019920.1 XP_016875409.1 6 - 6 3772 121
XM_017019921.1 XP_016875410.1 6 - 6 3235 121
XM_017019922.1 XP_016875411.1 5 - 5 4121 121
XM_017019923.1 XP_016875412.1 5 - 5 4052 121
GenBank mRNA AK000762.1 - 1 - 1 2252 121
AL833057.1 - 1 - 1 1826 121
BC008211.1 - 1 - 1 1789 121
BM718210.1 - 1 - 1 436 121
EU668356.1 - 4 - 4 2927 121

View Full Product Details