Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMTC1
Entrez Gene ID
83857
Gene Name
transmembrane and tetratricopeptide repeat containing 1
Gene Aliases
ARG99, OLF, TMTC1A
Chromosome Location
Chr.12: 29500813 - 29784759 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193451.1 NP_001180380.1 12 - 13 1945 82
NM_175861.3 NP_787057.2 12 - 13 2036 82
XM_005253499.4 XP_005253556.1 14 - 15 2197 82
XM_017020003.1 XP_016875492.1 13 - 14 2030 82
XM_017020004.1 XP_016875493.1 13 - 14 1990 82
XM_017020005.1 XP_016875494.1 11 - 12 1803 82
XM_017020007.1 XP_016875496.1 10 - 11 1223 82
GenBank mRNA AK055962.1 - 10 - 11 1197 82
AK123619.1 - 8 - 9 1627 82
BC042083.1 - 12 - 13 2036 82
GQ865601.1 - 12 - 13 1945 82

View Full Product Details