The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193451.1 | NP_001180380.1 | 3 - 4 | 615 | 82 |
NM_175861.3 | NP_787057.2 | 3 - 4 | 706 | 82 | |
XM_005253499.4 | XP_005253556.1 | 3 - 4 | 681 | 82 | |
XM_017020003.1 | XP_016875492.1 | 3 - 4 | 682 | 82 | |
XM_017020004.1 | XP_016875493.1 | 3 - 4 | 681 | 82 | |
XM_017020005.1 | XP_016875494.1 | 3 - 4 | 680 | 82 | |
XM_017020006.1 | XP_016875495.1 | 3 - 4 | 685 | 82 | |
GenBank mRNA | AK127297.1 | - | 3 - 4 | 491 | 82 |
BC042083.1 | - | 3 - 4 | 706 | 82 | |
GQ865601.1 | - | 3 - 4 | 615 | 82 |