Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPFIBP1
Entrez Gene ID
8496
Gene Name
PPFIA binding protein 1
Gene Aliases
L2, SGT2, hSGT2, hSgt2p
Chromosome Location
Chr.12: 27524112 - 27695564 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198915.1 NP_001185844.1 19 - 20 2131 78
NM_001198916.1 NP_001185845.1 20 - 21 2343 78
NM_003622.3 NP_003613.3 22 - 23 2418 78
NM_177444.2 NP_803193.2 21 - 22 2436 78
XM_005253505.1 XP_005253562.1 22 - 23 2214 78
XM_005253507.1 XP_005253564.1 22 - 23 2205 78
XM_005253508.1 XP_005253565.1 21 - 22 2184 78
XM_005253509.1 XP_005253566.1 21 - 22 2181 78
XM_006719159.3 XP_006719222.1 21 - 22 2168 78
XM_017020056.1 XP_016875545.1 24 - 25 2530 78
XM_017020057.1 XP_016875546.1 22 - 23 2705 78
XM_017020058.1 XP_016875547.1 21 - 22 2175 78
XM_017020059.1 XP_016875548.1 22 - 23 2464 78
XM_017020060.1 XP_016875549.1 20 - 21 2151 78
XM_017020061.1 XP_016875550.1 22 - 23 2438 78
XM_017020062.1 XP_016875551.1 21 - 22 2428 78
XM_017020063.1 XP_016875552.1 20 - 21 2401 78
XM_017020064.1 XP_016875553.1 22 - 23 2378 78
XM_017020065.1 XP_016875554.1 20 - 21 2375 78
XM_017020066.1 XP_016875555.1 21 - 22 2357 78
XM_017020067.1 XP_016875556.1 21 - 22 2354 78
XM_017020068.1 XP_016875557.1 21 - 22 2370 78
XM_017020069.1 XP_016875558.1 21 - 22 2345 78
XM_017020070.1 XP_016875559.1 21 - 22 2345 78
XM_017020071.1 XP_016875560.1 20 - 21 2324 78
XM_017020072.1 XP_016875561.1 20 - 21 2321 78
XM_017020073.1 XP_016875562.1 20 - 21 2332 78
XM_017020074.1 XP_016875563.1 19 - 20 2122 78
XM_017020075.1 XP_016875564.1 19 - 20 2020 78
XM_017020076.1 XP_016875565.1 12 - 13 1196 78
GenBank mRNA AB033056.2 - 19 - 20 2061 78
AF034802.1 - 22 - 23 2281 78
AK294267.1 - 18 - 19 1965 78
AK314350.1 - 20 - 21 2231 78
BC050281.1 - 21 - 22 2342 78
BX648761.1 - 4 - 5 750 78

View Full Product Details