Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9orf72
Entrez Gene ID
203228
Gene Name
chromosome 9 open reading frame 72
Gene Aliases
ALSFTD, FTDALS, FTDALS1
Chromosome Location
Chr.9: 27546546 - 27573866 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256054.2 NP_001242983.1 1 - 2 162 102
GenBank mRNA BQ068108.1 - - 173 102
JN681271.1 - 1 - 2 140 102

View Full Product Details