The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014705.3 | NP_055520.3 | 39 - 40 | 4403 | 72 |
XM_006716188.2 | XP_006716251.1 | 40 - 41 | 4417 | 72 | |
XM_006716189.2 | XP_006716252.1 | 40 - 41 | 4417 | 72 | |
XM_017012819.1 | XP_016868308.1 | 40 - 41 | 4790 | 72 | |
XM_017012820.1 | XP_016868309.1 | 39 - 40 | 4326 | 72 | |
XM_017012821.1 | XP_016868310.1 | 40 - 41 | 4344 | 72 | |
XM_017012822.1 | XP_016868311.1 | 40 - 41 | 4935 | 72 | |
XM_017012823.1 | XP_016868312.1 | 40 - 41 | 4344 | 72 | |
XM_017012824.1 | XP_016868313.1 | 39 - 40 | 4317 | 72 | |
XM_017012825.1 | XP_016868314.1 | 40 - 41 | 4344 | 72 | |
GenBank mRNA | AB018259.2 | - | 24 - 25 | 2516 | 72 |
AK091557.1 | - | 21 - 22 | 2231 | 72 | |
AK294722.1 | - | 9 - 10 | 860 | 72 | |
AK294820.1 | - | 9 - 10 | 1123 | 72 | |
AY233380.1 | - | 39 - 40 | 4134 | 72 | |
BC117688.2 | - | 39 - 40 | 4134 | 72 | |
BC117689.1 | - | 41 - 42 | 4269 | 72 |