Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAV20
Entrez Gene ID
28663
Gene Name
T cell receptor alpha variable 20
Gene Aliases
TCRAV20S1, TCRAV30S1
Chromosome Location
Chr.14: 22040663 - 22041153 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK127936.1 - 6 - 7 1120 73
BC025727.1 - 5 - 6 758 73
BX647165.1 - 4 - 5 635 73

Gene Symbol
TRAC
Entrez Gene ID
28755
Gene Name
T-cell receptor alpha constant
Gene Aliases
IMD7, TCRA, TRA, TRCA
Chromosome Location
Chr.14: 22547506 - 22552132 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA X02592.1 - 5 - 6 848 73

Gene Symbol
TRAJ17
Entrez Gene ID
28738
Gene Name
T cell receptor alpha joining 17
Gene Aliases
-
Chromosome Location
Chr.14: 22526844 - 22526906 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC035680.1 - 5 - 6 890 73

Gene Symbol
TRDV2
Entrez Gene ID
28517
Gene Name
T cell receptor delta variable 2
Gene Aliases
hDV102S1
Chromosome Location
Chr.14: 22422546 - 22423042 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC039714.1 - 5 - 6 824 73
BC106077.1 - 5 - 6 830 73

View Full Product Details