Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNRC6B
Entrez Gene ID
23112
Gene Name
trinucleotide repeat containing 6B
Gene Aliases
-
Chromosome Location
Chr.22: 40044817 - 40335808 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024843.1 NP_001020014.1 7 - 8 868 56
GenBank mRNA BC028626.2 - 7 - 8 801 56

View Full Product Details